Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CFAP298
Entrez Gene ID
535482
Gene Name
cilia and flagella associated protein 298
Gene Aliases
C1H21orf59
Chromosome Location
Chr.1: 2923955 - 2936283 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046407.1 NP_001039872.1 3 - 4 605 140
GenBank mRNA BC110223.1 - 3 - 4 605 140

View Full Product Details