Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FEM1A
Entrez Gene ID
516550
Gene Name
fem-1 homolog A
Gene Aliases
-
Chromosome Location
Chr.7: 19353551 - 19357132 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046226.1 NP_001039691.1 1 - 1 3349 80
GenBank mRNA BC114111.1 - 1 - 1 3349 80

View Full Product Details