Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PNMA2
Entrez Gene ID
540301
Gene Name
PNMA family member 2
Gene Aliases
-
Chromosome Location
Chr.8: 73748636 - 73756508 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046473.2 NP_001039938.1 1 - 2 101 99
GenBank mRNA BC112516.1 - 1 - 2 66 99
EE906439.1 - 1 - 2 101 99

View Full Product Details