Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
REX1BD
Entrez Gene ID
511085
Gene Name
required for excision 1-B domain containing
Gene Aliases
C19orf60, C7H19orf60
Chromosome Location
Chr.7: 4599488 - 4602278 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075490.2 NP_001068958.1 1 - 2 106 121
GenBank mRNA BC118476.1 - 1 - 2 107 121

View Full Product Details