Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BCL11A
Entrez Gene ID
538680
Gene Name
BAF chromatin remodeling complex subunit BCL11A
Gene Aliases
-
Chromosome Location
Chr.11: 43208356 - 43310317 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076121.2 NP_001069589.1 1 - 2 427 71
GenBank mRNA BC118227.1 - 1 - 2 285 71
LC016540.1 - 1 - 2 145 71

View Full Product Details