Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SINHCAF
Entrez Gene ID
538649
Gene Name
SIN3-HDAC complex associated factor
Gene Aliases
FAM60A
Chromosome Location
Chr.5: 78658640 - 78688174 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076116.1 NP_001069584.1 4 - 5 488 68
GenBank mRNA BC119933.1 - 4 - 5 488 68

View Full Product Details