Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TKFC
Entrez Gene ID
512373
Gene Name
triokinase and FMN cyclase
Gene Aliases
DAK
Chromosome Location
Chr.29: 39842535 - 39854640 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024524.3 NP_001019695.2 16 - 17 1876 81
GenBank mRNA BC126551.1 - 16 - 17 1877 81
BT021593.1 - 16 - 17 1883 81

View Full Product Details