Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SP1
Entrez Gene ID
540741
Gene Name
Sp1 transcription factor
Gene Aliases
-
Chromosome Location
Chr.5: 26598340 - 26642821 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078027.1 NP_001071495.1 3 - 4 1770 60
GenBank mRNA BC126621.1 - 3 - 4 1770 60

View Full Product Details