Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CHCHD10
Entrez Gene ID
615846
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 10
Gene Aliases
C17H22ORF16
Chromosome Location
Chr.17: 71226030 - 71227888 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083774.2 NP_001077243.1 3 - 4 477 71
GenBank mRNA BC134433.1 - 3 - 4 478 71
BC134453.1 - 3 - 4 481 71

View Full Product Details