Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
P3H2
Entrez Gene ID
511799
Gene Name
prolyl 3-hydroxylase 2
Gene Aliases
LEPREL1
Chromosome Location
Chr.1: 77130195 - 77309576 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100345.2 NP_001093815.1 1 - 2 639 107
GenBank mRNA BC148129.1 - 1 - 2 642 107

View Full Product Details