Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACSM2B
Entrez Gene ID
512227
Gene Name
acyl-CoA synthetase medium-chain family member 2B
Gene Aliases
ACSM2A
Chromosome Location
Chr.25: 18130586 - 18210919 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101952.2 NP_001095422.1 12 - 13 1665 78
GenBank mRNA BC151513.1 - 12 - 13 1666 78

View Full Product Details