Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHODL
Entrez Gene ID
613942
Gene Name
chondrolectin
Gene Aliases
-
Chromosome Location
Chr.1: 18712587 - 18736860 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101228.1 NP_001094698.1 1 - 2 448 88
GenBank mRNA BC147976.1 - 1 - 2 448 88

View Full Product Details