Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TLNRD1
Entrez Gene ID
616452
Gene Name
talin rod domain containing 1
Gene Aliases
MESDC1
Chromosome Location
Chr.21: 26839878 - 26841836 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101258.1 NP_001094728.1 1 - 1 1848 132
GenBank mRNA BC150077.1 - 1 - 1 1848 132

View Full Product Details