Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SHROOM1
Entrez Gene ID
615958
Gene Name
shroom family member 1
Gene Aliases
-
Chromosome Location
Chr.7: 44412468 - 44421072 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102294.1 NP_001095764.1 4 - 5 1782 60
GenBank mRNA BC151639.1 - 4 - 5 1782 60

View Full Product Details