Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NUDT10
Entrez Gene ID
616931
Gene Name
nudix (nucleoside diphosphate linked moiety X)-type motif 10
Gene Aliases
NUDT11
Chromosome Location
Chr.X: 88911182 - 88916867 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035488.3 NP_001030565.2 2 - 3 688 136
GenBank mRNA AW344479.1 - 1 - 2 235 136
BT021837.1 - 2 - 3 689 137

View Full Product Details