Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CD1B
Entrez Gene ID
509004
Gene Name
CD1b molecule
Gene Aliases
CD1B3
Chromosome Location
Chr.3: 11713130 - 11718199 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046021.1 NP_001039486.1 2 - 3 328 85
GenBank mRNA DQ192542.1 - 2 - 3 328 85

View Full Product Details