Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CD1A
Entrez Gene ID
781011
Gene Name
CD1a molecule
Gene Aliases
-
Chromosome Location
Chr.3: 11859207 - 11863100 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075382.2 NP_001068850.2 5 - 6 1087 158

View Full Product Details