Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATP23
Entrez Gene ID
514014
Gene Name
ATP23 metallopeptidase and ATP synthase assembly factor homolog
Gene Aliases
-
Chromosome Location
Chr.5: 55516050 - 55539042 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205490.1 NP_001192419.1 5 - 6 716 110

View Full Product Details