Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BRS3
Entrez Gene ID
539011
Gene Name
bombesin receptor subtype 3
Gene Aliases
-
Chromosome Location
Chr.X: 20090334 - 20094597 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205612.1 NP_001192541.1 2 - 3 786 92

View Full Product Details