Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NPHS2
Entrez Gene ID
537633
Gene Name
NPHS2 stomatin family member, podocin
Gene Aliases
-
Chromosome Location
Chr.16: 60789794 - 60808194 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206107.1 NP_001193036.1 7 - 8 872 61

View Full Product Details