Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UBE2E1
Entrez Gene ID
615599
Gene Name
ubiquitin conjugating enzyme E2 E1
Gene Aliases
-
Chromosome Location
Chr.27: 42078804 - 42139670 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206216.2 NP_001193145.1 5 - 6 600 82

View Full Product Details