Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CILP2
Entrez Gene ID
520750
Gene Name
cartilage intermediate layer protein 2
Gene Aliases
-
Chromosome Location
Chr.7: 3782380 - 3791397 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192528.1 NP_001179457.1 7 - 8 1194 95

View Full Product Details