Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PTH2
Entrez Gene ID
617680
Gene Name
parathyroid hormone 2
Gene Aliases
-
Chromosome Location
Chr.18: 55880924 - 55883485 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206281.1 NP_001193210.1 1 - 2 122 99

View Full Product Details