Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
BICD2
Entrez Gene ID
282447
Gene Name
BICD cargo adaptor 2
Gene Aliases
-
Chromosome Location
Chr.8: 84256025 - 84311938 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206475.1 NP_001193404.1 7 - 8 2497 67
GenBank mRNA CO887403.1 - 2 - 3 296 67

View Full Product Details