Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLPP4
Entrez Gene ID
529067
Gene Name
phospholipid phosphatase 4
Gene Aliases
PPAPDC1A
Chromosome Location
Chr.26: 40509729 - 40656785 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206550.1 NP_001193479.1 2 - 3 203 81

View Full Product Details