Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KCNK7
Entrez Gene ID
532497
Gene Name
potassium two pore domain channel subfamily K member 7
Gene Aliases
-
Chromosome Location
Chr.29: 43774318 - 43777103 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192725.1 NP_001179654.1 2 - 3 721 90

View Full Product Details