Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KLHL33
Entrez Gene ID
100141057
Gene Name
kelch like family member 33
Gene Aliases
-
Chromosome Location
Chr.10: 26692545 - 26702598 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166652.3 NP_001160124.1 3 - 4 1719 78

View Full Product Details