Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NR5A2
Entrez Gene ID
541305
Gene Name
nuclear receptor subfamily 5 group A member 2
Gene Aliases
-
Chromosome Location
Chr.16: 78828934 - 78957232 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206816.1 NP_001193745.1 6 - 7 1230 60

View Full Product Details