Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DCX
Entrez Gene ID
787968
Gene Name
doublecortin
Gene Aliases
-
Chromosome Location
Chr.X: 59464712 - 59872354 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206965.1 NP_001193894.1 4 - 5 962 68

View Full Product Details