Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COLGALT2
Entrez Gene ID
789150
Gene Name
collagen beta(1-O)galactosyltransferase 2
Gene Aliases
GLT25D2
Chromosome Location
Chr.16: 64972407 - 65091021 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191302.2 NP_001178231.1 11 - 12 1609 71

View Full Product Details