Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
N4BP1
Entrez Gene ID
540948
Gene Name
NEDD4 binding protein 1
Gene Aliases
-
Chromosome Location
Chr.18: 16967300 - 17021360 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191433.1 NP_001178362.1 6 - 7 2396 87
GenBank mRNA EE896108.1 - 3 - 4 261 87
GH319153.1 - 5 - 6 525 87

View Full Product Details