The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_174771.3 | NP_777196.1 | 9 - 10 | 442 | 109 |
GenBank mRNA | AF175558.1 | - | 9 - 10 | 422 | 109 |
BC134435.1 | - | 9 - 10 | 442 | 109 | |
BC149173.1 | - | 8 - 9 | 979 | 109 |