Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TNNI2
Entrez Gene ID
506434
Gene Name
troponin I2, fast skeletal type
Gene Aliases
-
Chromosome Location
Chr.29: 49645742 - 49648340 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192094.1 NP_001179023.1 6 - 7 355 67
GenBank mRNA BC142168.1 - 6 - 7 335 67
HQ697268.1 - 3 - 4 279 67

View Full Product Details