Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TLCD2
Entrez Gene ID
506954
Gene Name
TLC domain containing 2
Gene Aliases
-
Chromosome Location
Chr.19: 22802669 - 22808218 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192123.1 NP_001179052.1 3 - 4 448 72

View Full Product Details