Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FGF11
Entrez Gene ID
613386
Gene Name
fibroblast growth factor 11
Gene Aliases
-
Chromosome Location
Chr.19: 27115122 - 27121780 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192939.2 NP_001179868.1 3 - 4 410 90

View Full Product Details