Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SULT1C3
Entrez Gene ID
615528
Gene Name
sulfotransferase family, cytosolic, 1C, member 3
Gene Aliases
-
Chromosome Location
Chr.11: 44906839 - 44921512 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193080.1 NP_001180009.1 4 - 5 526 66

View Full Product Details