Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TCN1
Entrez Gene ID
616239
Gene Name
transcobalamin 1
Gene Aliases
-
Chromosome Location
Chr.15: 83058093 - 83073397 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193105.2 NP_001180034.1 3 - 4 409 91

View Full Product Details