Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PJVK
Entrez Gene ID
618114
Gene Name
pejvakin
Gene Aliases
DFNB59
Chromosome Location
Chr.2: 18429578 - 18437394 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193183.1 NP_001180112.1 5 - 6 769 88

View Full Product Details