The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014392.1 | NP_001014414.1 | 1 - 2 | 599 | 72 |
GenBank mRNA | AJ890840.1 | - | 1 - 2 | 599 | 72 |