Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C14H8orf34
Entrez Gene ID
615152
Gene Name
chromosome 14 C8orf34 homolog
Gene Aliases
C8orf34
Chromosome Location
Chr.14: 32378240 - 32742709 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199131.1 NP_001186060.1 8 - 9 1456 84

View Full Product Details