Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPIRE2
Entrez Gene ID
518761
Gene Name
spire type actin nucleation factor 2
Gene Aliases
-
Chromosome Location
Chr.18: 14652333 - 14679143 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205387.1 NP_001192316.1 8 - 9 1354 59
GenBank mRNA DV914571.1 - 2 - 3 292 59

View Full Product Details