Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VASN
Entrez Gene ID
787995
Gene Name
vasorin
Gene Aliases
SLITL2
Chromosome Location
Chr.25: 3526090 - 3536781 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083796.1 NP_001077265.1 2 - 2 2418 95
GenBank mRNA BC134480.1 - 2 - 2 2418 95

View Full Product Details