Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TGIF2
Entrez Gene ID
535632
Gene Name
TGFB induced factor homeobox 2
Gene Aliases
-
Chromosome Location
Chr.13: 65676251 - 65696116 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206085.3 NP_001193014.1 2 - 2 1818 74

View Full Product Details