Product Details

SNP ID
rs75582018
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:190428661 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAGTTTGGTCAAACTAAATTTTAG[A/T]TATCTATTGATGTTTATTGTTTTGT
Phenotype
MIM: 614786
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TMEM207 PubMed Links

Gene Details

Gene
TMEM207
Gene Name
transmembrane protein 207
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207316.2 1442 UTR 3 NP_997199.1

View Full Product Details