Product Details

SNP ID
rs77787064
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:7400759 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCTTTTTATGCATTTCTATACAC[A/G]CAGGAAATCTTACTCTGCAGGTGCT
Phenotype
MIM: 103071
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADCY2 PubMed Links

Gene Details

Gene
ADCY2
Gene Name
adenylate cyclase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020546.2 Intron NP_065433.2
XM_011513942.2 Intron XP_011512244.1

View Full Product Details