Product Details

SNP ID
rs78833648
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:42880985 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGGAGGTAATGGTGGGGAACCAA[C/G]AGAAAGTAATTAGAATTTTTATTTG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C6orf226 PubMed Links

Gene Details

Gene
C6orf226
Gene Name
chromosome 6 open reading frame 226
There are no transcripts associated with this gene.

Gene
RPL7L1
Gene Name
ribosomal protein L7 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198486.3 Intron NP_940888.3
XM_017010780.1 Intron XP_016866269.1

View Full Product Details