Product Details

SNP ID
rs79887583
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:141560380 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGATCTTACCCTGTAGTCTTCTTG[A/C]GTGGAAGCTGCCTTGTCCTTTCTTT
Phenotype
MIM: 610345
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
AGK PubMed Links

Gene Details

Gene
AGK
Gene Name
acylglycerol kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018238.3 Intron NP_060708.1
XM_005250023.3 Intron XP_005250080.1
XM_011516397.2 Intron XP_011514699.1

View Full Product Details