Product Details

SNP ID
rs75703315
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:92532521 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAAAAACTACAAATGCGGAAGGC[A/C]TATGTAGTAAAAACTACTGAAAATA
Phenotype
MIM: 602136
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PEX1 PubMed Links

Gene Details

Gene
PEX1
Gene Name
peroxisomal biogenesis factor 1
There are no transcripts associated with this gene.

Gene
RBM48
Gene Name
RNA binding motif protein 48
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032120.3 525 Silent Mutation GCA,GCC A140A NP_115496.2
XM_005250636.4 525 Silent Mutation GCA,GCC A140A XP_005250693.1
XM_006716149.3 525 Silent Mutation GCA,GCC A140A XP_006716212.1
XM_006716150.3 525 Silent Mutation GCA,GCC A140A XP_006716213.1
XM_017012709.1 525 UTR 5 XP_016868198.1

View Full Product Details