Product Details
- SNP ID
-
rs76603546
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:130733698 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCACGGGATCGGGTCGGAGAAGCC[A/G]GACGAGGCGCGTGGAGCTTCCCCCG
- Phenotype
-
MIM: 609393
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KLF14
PubMed Links
Gene Details
- Gene
- KLF14
- Gene Name
- Kruppel like factor 14
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_138693.3 |
336 |
Silent Mutation |
TCC,TCT |
S112S |
NP_619638.2 |
View Full Product Details