Product Details

SNP ID
rs78149791
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:35163623 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTTGGACTTTGCTGTGTCTCTTG[G/T]ACAGTTTAATTCTCTACGGCCACAC
Phenotype
MIM: 605836
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
UNC13B PubMed Links

Gene Details

Gene
UNC13B
Gene Name
unc-13 homolog B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006377.3 Intron NP_006368.3
XM_011517681.2 Intron XP_011515983.1
XM_011517682.2 Intron XP_011515984.1
XM_011517683.2 Intron XP_011515985.1
XM_011517684.2 Intron XP_011515986.1
XM_011517685.2 Intron XP_011515987.1
XM_011517686.2 Intron XP_011515988.1
XM_017014191.1 Intron XP_016869680.1
XM_017014192.1 Intron XP_016869681.1

View Full Product Details